missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p73 Antibody (5B1288), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56674DL594
This item is not returnable.
View return policy
Description
p73 Monoclonal antibody specifically detects p73 in Human, Mouse samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP assay (ChIP)
Specifications
| p73 | |
| Monoclonal | |
| DyLight 594 | |
| p53-related protein, P73p53-like transcription factor, TA p73, TAp73, tumor protein p73 | |
| This antibody was raised against full-length human p73. The epitope is thought to lie around the center of the molecule (NP_005418). | |
| 0.1 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 7161 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| 5B1288 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction