missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p73 Antibody (P73/2531), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08760IR
This item is not returnable.
View return policy
Description
p73 Monoclonal antibody specifically detects p73 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| p73 | |
| Monoclonal | |
| DyLight 755 | |
| p53-related protein, P73p53-like transcription factor, TA p73, TAp73, tumor protein p73 | |
| Recombinant full-length human p73 protein (Uniprot: O15350) | |
| 100 μg | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 7161 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| P73/2531 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction