missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p73 delta Monoclonal Antibody (DNp73-1.1)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA546190
This item is not returnable.
View return policy
Description
This antibody recognizes human deltaNp73; target epitope is MLYVGDPARHLAT.
p73 was identified as a long-lost cousin of tumor suppressor protein, p53 (4). p73 has high homology with p53 as well as with p63, a gene implicated in the maintenance of epithelial stem cells. Significant homology between p53, p63, and p73 (approximately 63% amino acid identity in the DNA-binding domain suggest that they may have overlapping functions in the regulation of gene expression (3). The targeted disruption of p73 gene leads to defects hippocampal dysgenesis, hydrocephalus, chronic inflammation and infections (2). Recently, spilicing variant mRNAs of p73 has been identified in MCF-7, a breast carcinoma cell line. These mRNAs code for variant p73 proteins bearing distinct carboxy-terminal structures suggesting that the carboxy-terminal region of p73 may be important for the functions of this protein (1) It was found p73 to be upregulated in a significant fraction of anaplastic thyroid cancers, whereas it was not detectable in normal thyroid epithelial cells nor in papillary or follicular thyroid cancer.
Specifications
| p73 delta | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| O15350 | |
| TP73 | |
| KLH-coupled synthetic peptide MLYVGDPARHLATC. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| DNp73-1.1 | |
| Unconjugated | |
| TP73 | |
| deltaNp73; OTTHUMP00000003479; OTTHUMP00000003480; p53-like transcription factor; p53-related protein; P73; RP23-254N4.10; TAp73; TP73; transformation related protein 73; Trp73; Tumor protein p73 | |
| Mouse | |
| Protein A | |
| RUO | |
| 7161 | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction