missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p73 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586461
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
P73 (TP73) participates in the apoptotic response to DNA damage. Isoforms containing the transactivation domain are pro-apoptotic, isoforms lacking the domain are anti-apoptotic and block the function of p53 and transactivating p73 isoforms. May be a tumor suppressor protein.
Specifications
| p73 | |
| Polyclonal | |
| Unconjugated | |
| TP73 | |
| deltaNp73; OTTHUMP00000003479; OTTHUMP00000003480; p53-like transcription factor; p53-related protein; P73; RP23-254N4.10; TAp73; Tp73; transformation related protein 73; Trp73; Tumor protein p73 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22062, 362675, 7161 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15350, Q9JJP2 | |
| TP73, Trp73 | |
| Synthetic peptide, corresponding to amino acids 271-320 of Human p73. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction