Learn More
Dostmann Electronic™ P755 Model 2-channel Multi-function Handheld Instrument
Click to view available options
Temperature Range (Metric):
-200°C to +850°C
Unit Size:
Each
Specifications
Specifications
| For Use With (Application) | Areas of application: For taking measurements for quality assurance under ISO 9000, As a reference instrument for checking production |
| Height (Metric) | 44 mm |
| Humidity Range | 0 to 100% |
| Length (Metric) | 200 mm |
| Model | P755 |
| Weight (Metric) | 300 g |
| Product Type | 2-channel Multi-function Handheld Instrument |
| Max. Temperature (Metric) | 850°C |
| Min. Temperature (Metric) | -200°C |
| Temperature Range (Metric) | -200°C to +850°C |
| Show More |