missing translation for 'onlineSavingsMsg'
Learn More

P7C3-A20, MedChemExpress

Product Code. p-200096131
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16446948

Brand: Medchem Express HY15978

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress P7C3-A20 is a derivative of P7C3 with potent proneurogenic and neuroprotective activity. P7C3-A20 exerts an antidepressant-like effect. P7C3-A20 can cross the blood-brain barrier and therefore has the potential for brain injury treatment.

P7C3-A20 is a derivative of P7C3 with potent proneurogenic and neuroprotective activity. P7C3-A20 exerts an antidepressant-like effect. P7C3-A20 can cross the blood-brain barrier and therefore has the potential for brain injury treatment.

Specifications

Chemical Name or Material P7C3-A20
CAS 1235481-90-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H19Br2FN2O
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (197.55 mM)
SMILES FC(CNC1=CC(OC)=CC=C1)CN2C3=CC=C(Br)C=C3C4=CC(Br)=CC=C24
Molecular Weight (g/mol) 506.21
Formula Weight 506.21
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.