missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPIL2 Antibody (015), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90282AF647
This item is not returnable.
View return policy
Description
PPIL2 Monoclonal specifically detects PPIL2 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| PPIL2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 23759 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunofluorescence | |
| 15 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| CYC4, cyclophilin, 60kDa, cyclophilin-60, Cyclophilin-like protein Cyp-60, Cyp-60, CYP60, EC 5.2.1.8, FLJ39930, hCyP-60, MGC33174, MGC787, peptidylprolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase-like 2, peptidylprolyl isomerase (cyclophilin)-like 2, PPIase, Rotamase PPIL2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human PPIL2 (Q13356-2; Gly280-Pro457). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction