missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
PAAF1 Antibody (OTI1F11), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73214B
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
PAAF1 Monoclonal specifically detects PAAF1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Tekniske data
| PAAF1 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| PAAF1 | |
| Full length recombinant protein of human PAAF1 (NP_079431) produced in HEK293T cells. | |
| 0.1 mL | |
| 80227.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI1F11 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| FLJ11848, PAAF, proteasomal ATPase-associated factor 1, Protein G-16, Rpn14, WD repeat domain 71, WD repeat-containing protein 71, WDR71 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse | |
| IgG2b |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion