Learn More
PAAF1 Mouse anti-Human, Clone: OTI1F11, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA507207
This item is not returnable.
View return policy
Description
PAAF1 inhibits proteasome 26S assembly and proteolytic activity by impairing the association of the 19S regulatory complex with the 20S core. In case of HIV-1 infection, recruited by viral Tat to the HIV-1 promoter, where it promotes the recruitment of 19S regulatory complex through dissociation of the proteasome 26S. This presumably promotes provirus transcription efficiency.Specifications
| PAAF1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BRP4 | |
| PAAF1 | |
| Full length human recombit protein of human PAAF1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1F11 | |
| Unconjugated | |
| PAAF1 | |
| PAAF, Rpn14, WDR71 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 80227 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |