missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PABPC1L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571307
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 86%.
PABPC1L is a protein coding gene. Gene Ontology (GO) annotations related to this gene include RNA binding.
Specifications
| PABPC1L | |
| Polyclonal | |
| Unconjugated | |
| PABPC1L | |
| 1810053B01Rik; C20orf119; dJ1069P2.3; embryonic poly(A) binding protein; embryonic poly(A)-binding protein; EPAB; LOW QUALITY PROTEIN: polyadenylate-binding protein 1-like; PABPC1L; PABPC1L1; poly(A) binding protein cytoplasmic 1 like; poly(A) binding protein, cytoplasmic 1-like; polyadenylate-binding protein 1-like; RGD1563142 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 80336 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q4VXU2 | |
| PABPC1L | |
| synthetic peptide directed towards the C-terminal region of human PABPC1L. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction