missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PABPC1L2A Polyclonal Antibody
GREENER_CHOICE

Product Code. 15767481
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15767481 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15767481 Supplier Invitrogen™ Supplier No. PA559629

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: KSHKEREAER GAWARQSTSA DVKDFEEDTD EEATLR Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 50%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PABPC1L2A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PABPC1L2A
Gene Accession No. Q5JQF8
Gene Alias PABPC1L2; Pabpc1l2a; PABPC1L2B; poly(A) binding protein cytoplasmic 1 like 2A; poly(A) binding protein, cytoplasmic 1-like 2A; poly(A) binding protein, cytoplasmic 1-like 2B; polyadenylate-binding protein 1-like 2; RBM32A; RBM32B; RGD1566367; RNA binding motif protein 32A; RNA binding motif protein 32B; RNA-binding motif protein 32; RNA-binding protein 32
Gene Symbols PABPC1L2A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PABPC1L2A. Recombinant protein control fragment (Product #RP-104326).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340529
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.