Learn More
Invitrogen™ PABPC3 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5104022
Product Code. 16630475
  • € 493.00 / 100µL
Description
Antibody detects endogenous levels of total PABPC3.
Messenger RNA stability and translation initiation are extensively under the control of poly(A)-binding proteins (PABP). See PABPC1 (MIM 604679) for background information.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PABPC3
Polyclonal
Unconjugated
PABPC3
4932702K14Rik; AI428050; PABP3; PABP-3; Pabpc3; Pabpc6; PABPL3; poly A binding protein, cytoplasmic 1 pseudogene; poly(A) binding protein cytoplasmic 3; poly(A) binding protein, cytoplasmic 3; poly(A) binding protein, cytoplasmic 6; poly(A)-binding protein 3; poly(A)-binding protein-like 3; polyadenylate-binding protein 3; RP11-165I9.5; testis PABP; Testis-specific poly(A)-binding protein; testis-specific poly(A)-binding protein (PABP); testis-specific poly(A)-binding protein 3; tPABP
Rabbit
Affinity chromatography
RUO
5042, 67543
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9H361
PABPC3, Pabpc6
A synthesized peptide derived from human PABPC3(Accession Q9H361), corresponding to amino acid residues T351-G401.
100 μL
Primary
Human, Mouse, Monkey