missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PABPC5 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA551359

Product Code. 15709320

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: EPNPAGKKKK YLKAALYVGD LDPDVTEDML YKKFRPAGPL RFTRICRDPV TRSPLGYGYV NFRFPADAEW ALNTMNFDLI NGKPFRLMWS QPDDRLRKSG VGNIFIKNLD KSIDNR Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

PABPC5 encodes a protein that binds to the polyA tail found at the 3' end of most eukaryotic mRNAs. It is thought to play a role in the regulation of mRNA metabolic processes in the cytoplasm. PABPC5 is located in a gene-poor region within the X-specific 13d-sY43 subinterval of the chromosome Xq21.3/Yp11.2 homology block. It is located close to translocation breakpoints associated with premature ovarian failure, and is therefore a potential candidate gene for this disorder.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PABPC5
Polyclonal
Unconjugated
PABPC5
C820015E17; PABP5; PABP-5; Pabpc5; poly A binding protein, cytoplasmic 5; poly A binding protein, cytoplasmic 5-like; poly(A) binding protein cytoplasmic 5; poly(A) binding protein, cytoplasmic 5; polyadenylate-binding protein 5; polyadenylate-binding protein 5-like
Rabbit
Antigen Affinity Chromatography
RUO
140886
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96DU9
PABPC5
Recombinant protein corresponding to Human PABPC5. Recombinant protein control fragment (Product #RP-91974).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.