Learn More
PABPC5 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25582-1-AP-20UL
 View more versions of this product
Product Code. 18066905
  • € 145.00 / 20µL
Description
The PABPC5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PABPC5. This antibody recognizes human antigen. The PABPC5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
PABPC5
350 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
PABP 5, PABP5, PABPC5, Poly(A) binding protein 5
20 μL
140886
Store at -20°C. Stable for one year after shipment.