Learn More
PABPC5 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25582-1-AP-20UL
This item is not returnable.
View return policy
Description
The PABPC5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PABPC5. This antibody recognizes human antigen. The PABPC5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PABPC5 | |
| 350 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| PABP 5, PABP5, PABPC5, Poly(A) binding protein 5 | |
| 20 μL | |
| 140886 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |