missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAC1R Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97421MFV500
This item is not returnable.
View return policy
Description
PAC1R Polyclonal antibody specifically detects PAC1R in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| PAC1R | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| adenylate cyclase activating polypeptide 1 (pituitary) receptor type 1, adenylate cyclase activating polypeptide 1 (pituitary) receptor type I, PACAP type I receptor, PACAP-R1, PACAP-R-1, PACAPRI, PACAPRPAC1, pituitary adenylate cyclase activating polypeptide 1 receptor type I Hiphop, pituitary adenylate cyclase-activating polypeptide type I receptor | |
| Produced in rabbits immunized with E. coli-derived Human PAC1R fragment. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, GPCR, Neuroscience, Neurotransmission, Signal Transduction | |
| 117 | |
| Store at 4C in the dark. | |
| IgG |
| Immunocytochemistry | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction