missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAC1R Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97421-100ul
This item is not returnable.
View return policy
Description
PAC1R Polyclonal antibody specifically detects PAC1R in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| PAC1R | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| adenylate cyclase activating polypeptide 1 (pituitary) receptor type 1, adenylate cyclase activating polypeptide 1 (pituitary) receptor type I, PACAP type I receptor, PACAP-R1, PACAP-R-1, PACAPRI, PACAPRPAC1, pituitary adenylate cyclase activating polypeptide 1 receptor type I Hiphop, pituitary adenylate cyclase-activating polypeptide type I receptor | |
| Produced in rabbits immunized with E. coli-derived Human PAC1R fragment. | |
| 100 μg | |
| Cancer, Cell Cycle and Replication, GPCR, Neuroscience, Neurotransmission, Signal Transduction | |
| 117 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction