Learn More
PACAP/ADCYAP1 Antibody (zelminemab) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-28879JF585
Product Code. 30177842
  • € 431.00 / 0.10mL
Description
PACAP/ADCYAP1 Monoclonal antibody specifically detects PACAP/ADCYAP1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PACAP/ADCYAP1
Monoclonal
Janelia Fluor 585
adenylate cyclase activating polypeptide 1 (pituitary), PACAPMGC126852, pituitary adenylate cyclase-activating polypeptide
PAC1
0.1 mL
Signal Transduction
116
Store at 4°C in the dark.
IgG1
Flow Cytometry, ELISA, Functional Assay
zelminemab
50mM Sodium Borate
Human
Protein A purified