missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PACAP/ADCYAP1 Antibody (zelminemab) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28879JF669
This item is not returnable.
View return policy
Description
PACAP/ADCYAP1 Monoclonal antibody specifically detects PACAP/ADCYAP1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| PACAP/ADCYAP1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| adenylate cyclase activating polypeptide 1 (pituitary), PACAPMGC126852, pituitary adenylate cyclase-activating polypeptide | |
| PAC1 | |
| 0.1 mL | |
| Signal Transduction | |
| 116 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| zelminemab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido