Learn More
Verenigde Kunststof Bedrijven™ U and M Range Plastic Bottles
Product Code. 10003754
Click to view available options
Capacity (Metric):
500 mL
1000 mL
1500 mL
2000 mL
2500 mL
Quantity:
60
72
90
132
135
180
216
Unit Size:
Pack of 132
Pack of 135
Pack of 180
Pack of 216
Pack of 60
Pack of 72
Pack of 90
Product Code. 10003754
Brand: Verenigde Kunststof Bedrijven™ 103730
Only null left
Add to basket
Add to basket
Provides solutions tailored to your precise requirements. Verenigde Kunststof Bedrijven™ U and M Range Plastic Bottles have a rectangular shape for comfortable handling, as well as an extensive labeling area. Caps not included; sold separately.
  • HDPE bottles
  • Food-grade
  • Rectangular shape
  • Comfortable customer friendly design
  • Extensive labelling area
  • Suitable for a wide variety of applications
  • Narrow and wide neck finish
Caps Not Included.
Specifications
Capacity (Metric) 500 mL
Color Natural
Dimensions (L x W x H) 88.6 x 88.6 x 108.5 mm
Material HDPE
Packaging Cardboard box
Quantity 216
Weight (Metric) 40 gr
Neck Finish DIN 80
Volume (Metric) Brim 675 ml
Mouth Wide
Shape Rectangular
Product Type Wide Mouth Bottle