Learn More
VACUUBRAND™ Package Rotary Vane Pump RZ 6 with Vacuum Gauge DCP 3000
GREENER_CHOICE
Product Code. 17841157
Click to view available options
Plug Type:
CEE
Unit Size:
Each
Product Code. 17841157
Brand: VACUUBRAND™ 20798150
Only null left
Add to basket
Add to basket
The rotary vane pump package RZ 6 with oil mist filter, butterfly valve VS 16 and vacuum gauge DCP 3000 offers great performance in a compact design - even with the gas ballast open, they enable a very good ultimate vacuum.
  • High flow rates even at vacuum levels approaching ultimate vacuum
  • High water vapor tolerance due to efficient gas ballast; very good ultimate vacuum even with gas ballast
  • Vacuum-tight at switch-off; external anti-suckback valve not needed
  • Large oil volume: Long intervals between oil changes
  • Ease of maintenance due to telescopic design
TRUSTED_SUSTAINABILITY
Specifications
Max. Vacuum 0.002 mbar
Plug Type CEE
Flow Rate 5.7 m³/hr.
Pump Type Rotary Vane Vacuum Pump
Enclosure Rating IP40
Applications Degassing, Lyophilization, Fine Vacuum Distillation and Drying Chambers
Maximum Pressure 1.1 bar
Voltage 230 V