Learn More
Cytiva Packing Tubes
€ 757.00 - € 1364.00
Product Code | Brand | For Use With | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Length (Metric) | Price | Quantity & Availability | ||||
12475730
|
Cytiva
28-9868-16 |
HiScale 16 Column | 200 mm |
€ 757.00
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
12465730
|
Cytiva
28-9868-15 |
HiScale 16 Column | 400 mm |
€ 972.00
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
11623234
|
Cytiva
28-9802-51 |
HiScale 50 Column | 200 mm |
€ 1364.00
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
11633234
|
Cytiva
28-9803-83 |
HiScale 26 Column | 200 mm |
€ 1019.00
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
||||