missing translation for 'onlineSavingsMsg'
Learn More

PACSIN1 Mouse anti-Chicken, Human, Mouse, Pig, Rabbit, Rat, Clone: 1B7B4, Proteintech

Product Code. 17731169 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17731169 150 μL 150µL
17741169 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17731169 Supplier Proteintech Supplier No. 681151IG150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PACSIN1 Monoclonal antibody specifically detects PACSIN1 in Chicken, Human, Mouse, Pig, Rabbit, Rat samples. It is validated for IHC, WB, ELISA

Specifications

Antigen PACSIN1
Applications Immunohistochemistry, Western Blot, ELISA
Classification Monoclonal
Clone 1B7B4
Concentration 1000 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Gene Accession No. BC050644
Host Species Mouse
Purification Method Protein G purification
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 29993
Target Species Chicken, Human, Mouse, Pig, Rabbit, Rat
Content And Storage Store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.