missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PACSIN1 Mouse anti-Chicken, Human, Mouse, Pig, Rabbit, Rat, Clone: 1B7B4, Proteintech
Shop All Proteintech ProductsDescription
PACSIN1 Monoclonal antibody specifically detects PACSIN1 in Chicken, Human, Mouse, Pig, Rabbit, Rat samples. It is validated for IHC, WB, ELISA
Specifications
Specifications
| Antigen | PACSIN1 |
| Applications | Immunohistochemistry, Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 1B7B4 |
| Concentration | 1000 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide and 50% glycerol pH 7.3. |
| Gene Accession No. | NM_001080432 |
| Host Species | Mouse |
| Purification Method | Protein G purification |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?