missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PACSIN2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10518-2-AP-20UL
This item is not returnable.
View return policy
Description
PACSIN 2, a 486 AA phosphoprotein is a novel member of the PACSIN family of cytoplasmic adapter proteins. It is an isoform of PACSIN with a CDC15 N-terminal domain, a C-terminal SH3 domain, 3 conserved regions specific to the PACSIN family, and 3 asn-pro-phe (NPF) motifs, which potentially bind to EH domains. This protein is known to display a ubiquitous expression pattern with the highest levels of expression in brain. Reports suggest that PACSIN 2 participates in the organization of the actin cytoskeleton, the regulation of vesicular traffic. It also participates in endocytic internalization and endocytic receptor recycling.Specifications
| PACSIN2 | |
| Polyclonal | |
| Unconjugated | |
| PACSIN2 | |
| PACSIN2, SDPII, Syndapin 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11252, 23970 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UNF0 | |
| PACSIN2 | |
| PACSIN2 Fusion Protein Ag0809 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction