missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PACSIN3 Antibody (OTI4F8), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73221MFV610
This item is not returnable.
View return policy
Description
PACSIN3 Monoclonal antibody specifically detects PACSIN3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| PACSIN3 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI4F8 | |
| Flow Cytometry | |
| Endophilin I, FLJ17861, protein kinase C and casein kinase substrate in neurons 3, protein kinase C and casein kinase substrate in neurons protein 3, SDPIII, SH3 domain-containing protein 6511, syndapin III | |
| Full length human recombinant protein of human PACSIN3 (NP_057307) produced in HEK293T cell. | |
| 0.1 mL | |
| Cancer, Diabetes Research, Neuroscience, Protein Kinase, Signal Transduction, Transcription Factors and Regulators | |
| 29763 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction