missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PACSIN3 Monoclonal Antibody (OTI2A3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502251S
This item is not returnable.
View return policy
Description
PACSIN3 Monoclonal antibody specifically detects PACSIN3 in Canine, Human, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| PACSIN3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UKS6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 29763, 311187, 475984 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI2A3 | |
| Unconjugated | |
| PACSIN3 | |
| 4921507A02Rik; 6330413E15Rik; AW413130; EGK_06300; Endophilin I; PACSIN3; Protein kinase C and casein kinase II substrate protein 3; protein kinase C and casein kinase substrate in neurons 3; protein kinase C and casein kinase substrate in neurons protein 3; SDPIII; SH3 domain-containing protein 6511; syndapin III | |
| Full length human recombinant protein of human PACSIN3 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction