missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97680-100ul
This item is not returnable.
View return policy
Description
PAD3 Polyclonal antibody specifically detects PAD3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PAD3 | |
| Polyclonal | |
| Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| EC 3.5.3.15, MGC126307, PAD3, PDI3MGC126308, peptidyl arginine deiminase, type III, Peptidylarginine deiminase III, Protein-arginine deiminase type III, protein-arginine deiminase type-3 | |
| Produced in rabbits immunized with E. coli-derived Human PAD3 fragment | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 51702 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction