missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PADI2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93929-0.02ml
This item is not returnable.
View return policy
Description
PADI2 Polyclonal antibody specifically detects PADI2 in Human samples. It is validated for Western Blot
Specifications
| PADI2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EC 3.5.3.15, KIAA0994PDI2PAD2, PAD-H19, peptidlyarginine deiminase type II, peptidyl arginine deiminase, type II, Peptidylarginine deiminase II, protein arginine deiminase, Protein-arginine deiminase type II, protein-arginine deiminase type-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human PADI2 (NP_031391.2). MLRERTVRLQYGSRVEAVYVLGTYLWTDVYSAAPAGAQTFSLKHSEHVWVEVVRDGEAEEVATNGKQRWLLSPSTTLRVTMSQASTEASSDKVTVNYYDEEGSIPIDQAGLFLTAIEISLDVDADRDGVVEKNNPKKASWTWGPEGQGAILLVNCDRETPWLPKEDCRDEKVYSKEDLKDMSQMILRTKGPDRLPAGYEI | |
| 0.02 mL | |
| Alzheimers Research, Amino Acids Drugs and other small molecules, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 11240 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction