missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PADI3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
€ 181.00 - € 453.00
Specifications
| Antigen | PADI3 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17713114
|
Invitrogen™
PA5121713 |
20 μL |
€ 181.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17703114
![]() |
Invitrogen™
PA5121712 |
100 μL |
€ 453.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PADI3 Polyclonal antibody specifically detects PADI3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
| PADI3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9ULW8 | |
| 51702 | |
| Met1-Pro300 (Accession Q9ULW8). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| PADI3 |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| PAD3; PADI3; PDI3; peptidyl arginine deiminase 3; peptidyl arginine deiminase, type III; peptidylarginine deiminase III; Protein-arginine deiminase type III; protein-arginine deiminase type-3 | |
| PADI3 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title