Learn More
PADI4 Mouse anti-Human, Clone: OTI4H5, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF504813
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Catalyzes the citrullination/deimination of arginine residues of proteins. Citrullinates histone H3 at ′Arg-8′ and/or ′Arg-17′ and histone H4 at ′Arg-3′, which prevents their methylation by CARM1 and HRMT1L2/PRMT1 and represses transcription. Citrullinates EP300/P300 at ′Arg-2142′, which favors its interaction with NCOA2/GRIP1.Specifications
| PADI4 | |
| Monoclonal | |
| Unconjugated | |
| PADI4 | |
| PAD, PAD4, PADI5, PDI4, PDI5 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23569 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI4H5 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9UM07 | |
| PADI4 | |
| Full length human recombit protein of human PADI4 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.