missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAF1 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-273C
This item is not returnable.
View return policy
Description
PAF1 Polyclonal specifically detects PAF1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| PAF1 | |
| Polyclonal | |
| Western Blot | |
| F23149_1, FLJ11123, Paf1, RNA polymerase II associated factor, homolog (S. cerevisiae), Pancreatic differentiation protein 2, PD2hPAF1, RNA polymerase II-associated factor 1 homolog | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specific for human Paf1 protein. | |
| Store at 4C in the dark. |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PAF1 | |
| The immunogen recognized by this antibody maps to a region between residues 75 and 125 of human RNA Polymerase Associated Factor using the numbering given in TrEMBL entry Q9NUU9 (GeneID 54623). | |
| 100 μL | |
| Transcription Factors and Regulators | |
| 54623 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction