missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAF49 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583573
This item is not returnable.
View return policy
Description
Immunogen sequence: GNPPVTGPRS ALAPNLLTSG KKKKEMQVTE APVTQEAVNG HGALEVDMAL GSPEMDVRKK KKKKNQQLKE PEAAGPVGTE PTVETLEPLG VLFPSTTKKR KKPKGK Highest antigen sequence indentity to the following orthologs: Mouse - 44%, Rat - 47%.
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase I which synthesizes ribosomal RNA precursors. Isoform 1 is involved in UBTF-activated transcription, presumably at a step following PIC formation.; Isoform 2 has been described as a component of preformed T-cell receptor (TCR) complex. [UniProt].
Specifications
| PAF49 | |
| Polyclonal | |
| Unconjugated | |
| Cd3eap | |
| 1190028F09; 2610103M17Rik; A34.5; antis; Anti-sense to ERCC-1 protein; antisense to ERCC-1 protein; Ase1; ASE-1; CAST; CD3 epsilon associated protein; antisense to Ercc1; CD3E antigen, epsilon polypeptide associated protein; CD3E antigen, epsilon polypeptide-associated protein; CD3e molecule associated protein; CD3e molecule, epsilon associated protein; Cd3eap; CD3E-associated protein; CD3-epsilon-associated protein; DNA-directed RNA polymerase I subunit RPA34; PAF49; RGD1305916; RNA polymerase I-associated factor PAF49 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10849 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15446 | |
| Cd3eap | |
| Recombinant protein corresponding to Human CD3EAP. Recombinant protein control fragment (Product #RP-98992). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction