missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAFAH Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15526-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
PLA2G7 encodes a protein that is a secreted enzyme that catalyzes the degradation of platelet-activating factor to biologically inactive products. Defects in this gene are a cause of platelet-activating factor acetylhydrolase deficiency.Specifica
| PAFAH | |
| Polyclonal | |
| Unconjugated | |
| PLA2G7 | |
| Group VIIA phospholipase A2, gVIIA PLA2, LDL PLA(2), LDL PLA2, Lp-PLA2, PAF 2 acylhydrolase, PAF acetylhydrolase, PAFAH, PAF-AH, PLA2G7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 27226, 7941 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13093, Q60963 | |
| Pla2g7 | |
| PAFAH Fusion Protein Ag7227 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto