missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAG1 Recombinant Rabbit Monoclonal Antibody (24GB685)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557150
This item is not returnable.
View return policy
Description
PAG1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
PAG (phosphoprotein associated with GEMs), also known as Cbp (Csk-binding protein), is a ubiquitously expressed 46 kDa transmembrane adaptor protein present in membrane rafts (glycosphingolipid-enriched microdomains), which however migrates on SDS PAGE gels anomalously as an 80 kDa molecule. Following tyrosine phosphorylation by Src family kinases, PAG binds and thereby activates the protein tyrosine kinase Csk, the major negative regulator of the Src family kinases. Signaling via the B-cell receptor in B cells or high affinity IgE receptor (FcepsilonRI) in mast cells leads to PAG increased tyrosine phosphorylation and Csk binding, while T cell receptor signaling causes PAG dephosphorylation, loss of Csk binding and increased activation of the protein tyrosine kinase Lck.
Specifications
| PAG1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PAG1 | |
| CBP; Csk binding protein; csk-binding protein; F730007C19Rik; Pag; PAG 1; Pag1; PAG1B; phosphoprotein associated with glycosphingolipid microdomains 1; phosphoprotein associated with glycosphingolipid-enriched microdomains 1; phosphoprotein membrane anchor with glycosphingolipid microdomains 1; phosphoprotein transmembrane adaptor 1; phosphoprotein-associated with GEMs; pregnancy-associated glycoprotein 1; Pregnancy-specific protein B; PSP-B; similar to pregnancy-associated glycoprotein 1; transmembrane adapter protein PAG; transmembrane adaptor protein PAG; transmembrane phosphoprotein Cbp | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55824, 64019, 94212 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB685 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q3U1F9, Q9JM80, Q9NWQ8 | |
| PAG1 | |
| A synthesized peptide derived from human PAG1 (aa 400-432). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction