missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAGE1 Antibody (OTI3E7), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73230AF532
This item is not returnable.
View return policy
Description
PAGE1 Monoclonal specifically detects PAGE1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PAGE1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PAGE1 | |
| Full length recombinant protein of human PAGE1 (NP_003776) produced in E.coli. | |
| 0.1 mL | |
| 8712.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI3E7 | |
| Western Blot, Immunohistochemistry | |
| CT16.3, G antigen, family B, 1 (prostate associated), GAGE9, GAGE-9AL5, GAGEB1G antigen family B member 1, P antigen family, member 1 (prostate associated), PAGE-1G antigen 9, prostate associated gene 1, Prostate-associated gene 1 protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction