Learn More
PAGE1 Mouse anti-Human, Clone: OTI13B1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF805801
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
PAGE1 belongs to a family that are expressed in a variety of tumors but not in normal tissues, except for the testis. Nothing is presently known about the function of this protein.This gene belongs to a family of genes that are expressed in a variety of tumors but not in normal tissues, except for the testis. Unlike the other gene family members, this gene does not encode an antigenic peptide. Nothing is presently known about the function of this protein.Specifications
| PAGE1 | |
| Monoclonal | |
| Unconjugated | |
| PAGE1 | |
| AL5, CT16.3, GAGE-9, GAGEB1, PAGE-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8712 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI13B1 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O75459 | |
| PAGE1 | |
| Full length human recombit protein of human PAGE1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.