missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAH Monoclonal Antibody (717CT21.1.1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537648
This item is not returnable.
View return policy
Description
PAH Monoclonal Antibody for Western Blot, IHC (P)
PAH encodes the enzyme phenylalanine hydroxylase that is the rate-limiting step in phenylalanine catabolism. Deficiency of this enzyme activity results in the autosomal recessive disorder phenylketonuria.
Specifications
| PAH | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P00439, P04176, P16331 | |
| PAH | |
| KLH conjugated synthetic peptide between 127-161 amino acids from the Central region of human PAH. | |
| 400 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| 717CT21.1.1 | |
| Unconjugated | |
| PAH | |
| AW106920; Pah; PH; phe-4-monooxygenase; phenylalanine 4-monooxygenase; phenylalanine hydroxylase; phenylalanine-4-hydroxylase; PKU; PKU1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 18478, 24616, 5053 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction