Learn More
Invitrogen™ PAH Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5102422
Product Code. 16668894
  • € 493.00 / 100µL
Description
Antibody detects endogenous levels of total PAH.
PAH encodes the enzyme phenylalanine hydroxylase that is the rate-limiting step in phenylalanine catabolism. Deficiency of this enzyme activity results in the autosomal recessive disorder phenylketonuria.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PAH
Polyclonal
Unconjugated
PAH
AW106920; Pah; PH; phe-4-monooxygenase; phenylalanine 4-monooxygenase; phenylalanine hydroxylase; phenylalanine-4-hydroxylase; PKU; PKU1
Rabbit
Affinity chromatography
RUO
18478, 24616, 5053
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P00439, P04176, P16331
PAH
A synthesized peptide derived from human PAH(Accession P00439), corresponding to amino acid residues G46-K96.
100 μL
Primary
Human, Mouse, Rat