Learn More
Invitrogen™ PAH Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102422
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PAH.
PAH encodes the enzyme phenylalanine hydroxylase that is the rate-limiting step in phenylalanine catabolism. Deficiency of this enzyme activity results in the autosomal recessive disorder phenylketonuria.
Specifications
| PAH | |
| Polyclonal | |
| Unconjugated | |
| PAH | |
| AW106920; Pah; PH; phe-4-monooxygenase; phenylalanine 4-monooxygenase; phenylalanine hydroxylase; phenylalanine-4-hydroxylase; PKU; PKU1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18478, 24616, 5053 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P00439, P04176, P16331 | |
| PAH | |
| A synthesized peptide derived from human PAH(Accession P00439), corresponding to amino acid residues G46-K96. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |