Learn More
Paichongding, TRC
Specifications
Specifications
| Chemical Name or Material | Paichongding |
| CAS | 948994-16-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17H23ClN4O3 |
| Quantity | 100 mg |
| Synonym | 1-[(6-Chloro-3-pyridinyl)methyl]-1,2,3,5,6,7-hexahydro-7-methyl-8-nitro-5-propoxyimidazo[1,2-a]pyridine, 1-[(6-Chloropyridin-3-yl)-methyl]-5-propoxy-7-methyl-8-nitro-1,2,3,5,6,7-hexahydroimidazo[1,2-a]pyridine, 1-[(6-Chloropyridin-3-yl)methyl]-7-methyl-8-nitro-5-propoxy-1,2,3,5,6,7-hexahydroimidazo[1,2-a]pyridine, IPP, IPPA 152004, Paichongding |
| InChI Formula | InChI=1S/C17H23ClN4O3/c1-3-8-25-15-9-12(2)16(22(23)24)17-20(6-7-21(15)17)11-13-4-5-14(18)19-10-13/h4-5,10,12,15H,3,6-9,11H2,1-2H3 |
| SMILES | CCCOC1CC(C)C(=C2N(Cc3ccc(Cl)nc3)CCN12)[N+](=O)[O-] |
| IUPAC Name | 1-[(6-chloropyridin-3-yl)methyl]-7-methyl-8-nitro-5-propoxy-3,5,6,7-tetrahydro-2H-imidazo[1,2-a]pyridine |
| Molecular Weight (g/mol) | 366.84 |
| Show More |