missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAICS, Mouse anti-Human, Clone: 4F4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00010606-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MATAEVLNIGKKLYEGKTKEVYELLDSPGKVLLQSKDQITAGNAARKNHLEGKAAISNKITSCIFQLLQEAGIKTAFTRKCGETAFIAPQCEMIPIEWVCRRIATGSFLKRNPGVKEGYKFYPPKVELFFKDDANNDPQWSEEQLIAAKFCFAGLLIGQTEVDIMSHATQAIFEILEKSWLPQNCTLVDMKIEFGVDVTTKEIVLADVIDNDSWRLWPSGDRSQQKDKQSYRDLKEVTPEGLQMVKKNFEWVAERVELLLKSESQCRVVVLMGSTSDLGHCEKIKKACGNFGIPCELRVTSAHKGPDETLRIKAEYEGDGIPTVFVAVAGRSNGLGPVMSGNTAYPVISCPPLTPDWGVQDVWSSLRLPSGLGCSTVLSPEGSAQFAAQIFGLSNHLVWSKLRASILNTWISLKQADKKIRECNLSpecifications
| PAICS | |
| Monoclonal | |
| Unconjugated | |
| phosphoribosylaminoimidazole carboxylase, phosphoribosylaminoimidazole succinocarboxamide synthetase | |
| ADE2/ADE2H1/AIRC/DKFZp781N1372/MGC1343/MGC5024/PAIS | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 4F4 | |
| In 1x PBS, pH 7.4 | |
| BC010273 | |
| PAICS | |
| PAICS (AAH10273, 1 a.a. ∼ 425 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10606 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction