missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530394
This item is not returnable.
View return policy
Description
Recommended positive controls: A431. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (81%), Chicken (87%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene interacts with poly(A)-binding protein and with the cap-binding complex eIF4A. It is involved in translational initiation and protein biosynthesis. Overexpression of this gene in COS7 cells stimulates translation. Alternative splicing occurs at this locus and three transcript variants encoding three distinct isoforms have been identified.
Specifications
| PAIP1 | |
| Polyclonal | |
| Unconjugated | |
| PAIP1 | |
| PABC1-interacting protein 1; PABP-interacting protein 1; Paip1; PAIP-1; poly(A) binding protein interacting protein 1; poly(A)-binding protein-interacting protein 1; polyadenylate binding protein-interacting protein 1; polyadenylate-binding protein-interacting protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10605 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.79 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9H074 | |
| PAIP1 | |
| Recombinant protein encompassing a sequence within the center region of human PAIP1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction