missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAIP2 Antibody (1C12), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22555FR
This item is not returnable.
View return policy
Description
PAIP2 Monoclonal antibody specifically detects PAIP2 in Human samples. It is validated for Western Blot, ELISA
Specifications
| PAIP2 | |
| Monoclonal | |
| DyLight 680 | |
| MGC72018, PABC1-interacting protein 2, PABP-interacting protein 2, PAIP-2, PAIP2APoly(A)-binding protein-interacting protein 2, poly(A) binding protein interacting protein 2, polyA-binding protein-interacting protein 2, polyadenylate-binding protein-interacting protein 2 | |
| Recombinant human PAIP2 (1-127aa) purified from E. coli | |
| 0.1 mL | |
| metabolism | |
| 51247 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, ELISA | |
| 1C12 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction