missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PAK1, GST Tag Recombinant Protein

Product Code. 15428583
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
This item is not returnable. View return policy

Product Code. 15428583

Brand: Invitrogen™ PR7402A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length

PAK1 is a serine/threonine kinase that is a target for restenosis, cancer, and immunosuppression treatment. These proteins serve as targets for the small GTP binding proteins Cdc42 and Rac and have been implicated in a wide range of biological activities. PAK1 regulates cell motility and morphology.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13153
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 5058
Molecular Weight (g/mol) 88.1 kDa
Name Human PAK1, GST Tag
Quantity 100 μg
Regulatory Status RUO - research use only
Gene Alias activated protein kinase alpha; ADRB2; adriamycin resistance-associated; alpha pak; alpha-PAK; ARA; AW045634; Beta-PAK; bPAK; CDC42/RAC effector kinase PAK-A; C-t-PAK2; EC 2.7.11.1; gamma-PAK; kinase PAK1; MGC130000; MGC130001; MRX30; MRX47; Oligophrenin-3; OPHN3; p21 (CDKN1A)-activated kinase 1; p21 (CDKN1A)-activated kinase 2; p21 (CDKN1A)-activated kinase 3; p21 (RAC1) activated kinase 1; p21 (RAC1) activated kinase 2; p21 (RAC1) activated kinase 3; p21 protein (Cdc42/Rac)-activated kinase 1; p21 protein (Cdc42/Rac)-activated kinase 2; p21 protein (Cdc42/Rac)-activated kinase 3; p21/Cdc42/Rac1-activated kinase 1 (STE20 homolog, yeast); p21/Cdc42/Rac1-activated kinase 1 (yeast Ste20-related); p21-activated kinase 1; p21-activated kinase 2; p21-activated kinase 3; p27; p34; p58; p65-PAK; p68-PAK; PAK 1; PAK1; PAK-1; PAK1B; PAK2; PAK-2; PAK-2p27; PAK-2p34; PAK3; PAK-3; PAK3beta; PAK65; Paka; PAKalpha; PAKgamma; protein kinase MUK2; S6/H4 kinase; serine/threonine-protein kinase PAK 1; Serine/threonine-protein kinase PAK 2; serine/threonine-protein kinase PAK 3; STE20 homolog, yeast
Common Name PAK1
KinaseFamily STE20 Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.