missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAK3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19682
This item is not returnable.
View return policy
Description
PAK3 Polyclonal specifically detects PAK3 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| PAK3 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| PAK3 | |
| Recombinant protein encompassing a sequence within the center region of human PAK3. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse. Species reactivity based on sequence identity: Rat (100%), Xenopus laevis (100%), Bovine (100%), Chimpanzee (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.74mg/mL | |
| Western Blot 1:500-1:3000 | |
| beta-PAK, bPAK, CDKN1A, EC 2.7.11, EC 2.7.11.1, hPAK3, mental retardation, X-linked 47, MRX30, MRX47, Oligophrenin-3, OPHN3, p21 (CDKN1A)-activated kinase 3, p21 protein (Cdc42/Rac)-activated kinase 3, p21-activated kinase 3, PAK-3, PAK3beta, serine/threonine-protein kinase PAK 3 | |
| Rabbit | |
| 62 kDa | |
| 0.1 mL | |
| Protein Kinase | |
| 5063 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction