missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAK4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19683
This item is not returnable.
View return policy
Description
PAK4 Polyclonal antibody specifically detects PAK4 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| PAK4 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| EC 2.7.11, EC 2.7.11.1, KIAA1142, p21 protein (Cdc42/Rac)-activated kinase 4, p21(CDKN1A)-activated kinase 4, p21-activated kinase 4, PAK-4, protein kinase related to S. cerevisiae STE20, effector for Cdc42Hs, serine/threonine-protein kinase PAK 4 | |
| Rabbit | |
| 64 kDa | |
| 0.1 mL | |
| Apoptosis, Protein Kinase | |
| 10298 | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| O96013 | |
| PAK4 | |
| Recombinant protein encompassing a sequence within the center region of human PAK4. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Bovine (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction