Learn More
Thermo Scientific Chemicals Palatinose hydrate, 98+%
Product Code. 15412929
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15412929 100 g 100g
15402929 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15412929 Supplier Thermo Scientific Chemicals Supplier No. J60091.22
Request Bulk or Custom Format
CAS: 343336-76-5 | C12H22O11 | 342.30 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 343336-76-5
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00076094
InChI Key PVXPPJIGRGXGCY-VVLSOAGWNA-N
Synonym 6-o-alpha-d-glucopyranosyl-beta-d-fructofuranose, beta-palatinose, beta-f-palatinose, alpha-d-glucopyranose-1->6-beta-d-fructofuranose, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2r,3s,4s,5r-3,4,5-trihydroxy-5-hydroxymethyl oxolan-2-yl methoxy oxane-3,4,5-triol
PubChem CID 52994241
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[[(2R,3S,4S)-3,4,5-trihydroxy-5-(hydroxymethyl)oxolan-2-yl]methoxy]oxane-3,4,5-triol;hydrate
SMILES OC[C@H]1O[C@H](OC[C@H]2O[C@@](O)(CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
Specifications
Melting Point 122°C to 124°C
Color White
Quantity 100 g
Beilstein 1757215
Merck Index 14,5182
Formula Weight 342.3
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Palatinose hydrate