missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PALB2 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-60439AF488
This item is not returnable.
View return policy
Description
PALB2 Polyclonal antibody specifically detects PALB2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| PALB2 | |
| Alexa Fluor 488 | |
| DKFZp667I166, DKFZp686E1054, FANCNFLJ21816, partner and localizer of BRCA2, PNCA3 | |
| 0.1 mL | |
| Mouse | |
| Purified |
| Polyclonal | |
| 50mM Sodium Borate | |
| Immunogen affinity purified | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction