missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PALB2 Antibody (M41-P3F3), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50232G
This item is not returnable.
View return policy
Description
PALB2 Monoclonal specifically detects PALB2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| PALB2 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PALB2 | |
| Peptide Sequence - KENQFLMPPEE | |
| 0.1 mL | |
| Breast Cancer, Cancer | |
| 79728 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry | |
| M41-P3F3 | |
| Western Blot, ELISA, Immunohistochemistry | |
| DKFZp667I166, DKFZp686E1054, FANCNFLJ21816, partner and localizer of BRCA2, PNCA3 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction