Learn More
Palladium(II) 2,2,2-trifluoroacetate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 42196-31-6 |
| Molecular Formula | C4F6O4Pd |
| MDL Number | MFCD00013204 |
| Quantity | 5 g |
| SMILES | O=C([O-])C(F)(F)F.O=C([O-])C(F)(F)F.[Pd+2] |
| Molecular Weight (g/mol) | 332.45 |
| Percent Purity | 98% |
| Chemical Name or Material | Palladium(II) 2,2,2-trifluoroacetate |