Learn More
Invitrogen™ PALM2-AKAP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140342
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: VLGYDETIKA ELVLIDEDDE KSLREKTVTD VSTIDGNAAE LVSGRPVSDT TEPSSPEGKE ES Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 97%.
Specifications
| PALM2-AKAP2 | |
| Polyclonal | |
| Unconjugated | |
| PALM2 | |
| A kinase (PRKA) anchor protein 2; AKAP2; AKAP-2; AKAPKL; A-kinase anchor protein 2; A-kinase anchoring protein 2; MISP2; PALM2; paralemmin 2; paralemmin-2; PRKA2; protein kinase A anchoring protein 2; protein kinase A2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11217, 114299 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IXS6, Q9Y2D5 | |
| AKAP2, PALM2 | |
| Recombinant protein corresponding to Human PALM2AKAP2. Recombinant protein control fragment (Product #RP-109594). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.